Febuxostat ≥97.0% (by HPLC, titration analysis)

Leverantör: TCI
Varning

Synonyms: 2-(3-Cyano-4-isobutoxyphenyl)-4-methylthiazole-5-carboxylic acid

F0847-1G F0847-5G
TCIAF0847-1GEA 230 SEK
TCIAF0847-1G TCIAF0847-5G
Febuxostat ≥97.0% (by HPLC, titration analysis)
Febuxostat
Formel: C₁₆H₁₆N₂O₃S
MDL Number: MFCD00871598
CAS nummer: 144060-53-7
Merck Index: 14,03948

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (Neutralization titration) min. 97.0 %
Melting point 207.0 to 211.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR