Florfenicol ≥98.0% (by HPLC, total nitrogen)
Leverantör: TCI
Synonyms:
2,2-Dichloro-N-[(1R,2S)-3-fluoro-1-hydroxy-1-(4-methylsulphonylphenyl)-2-propyl]acetamide, D-(-)-threo-2-Dichloroacetamido-3-fluoro-1-(4-methylsulphonylphenyl)-1-propanol, 2,2-Dichloro-N-((1R,2S)-3-fluoro-1-hydroxy-1-(4-(methylsulphonyl)phenyl)propan-2-yl)acetamide
F0811-1G
F0811-5G
TCIAF0811-1GEA
710
SEK
TCIAF0811-1G
TCIAF0811-5G
Florfenicol ≥98.0% (by HPLC, total nitrogen)
Florfenicol
Formel:
C₁₂H₁₄Cl₂FNO₄S MW: 358.21 g/mol |
MDL Number:
MFCD00864834 CAS nummer: 73231-34-2 Merck Index: 14,04109 |
Specifikation testresultat
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Melting point | 153.0 to 157.0 deg-C |
Specific rotation [a]20/D | -16.0 to -19.0 deg (C=1, DMF) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...