Fmoc-Arg(Pbf)-OH 98,0 HPLC, T
Leverantör: TCI
Synonyms: Nα-Fmoc-Nω-Pbf-L-arginine, Nα-Fmoc-Nomega-(2,2,4,6,7-pentamethyl-2,3-dihydrobenzo[b]furan-5-ylsulphonyl)-L-arginine, Nɑ-[(9H-Fluoren-9-ylmethoxy)carbonyl]-Nω-(2,2,4,6,7-pentamethyldihydrobenzofuran-5-sulphonyl)-L-arginine
F0729-5G F0729-25G
TCIAF0729-5GEA 725 SEK
TCIAF0729-5G TCIAF0729-25G
Fmoc-Arg(Pbf)-OH 98,0 HPLC, T
Fmoc-Arg(Pbf)-OH
Formel: C₃₄H₄₀N₄O₇S
MW: 648.78 g/mol
Storage Temperature: Frys
MDL Number: MFCD00235804
CAS nummer: 154445-77-9
Order Now
Specifikation testresultat
Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Specific rotation [a]20/D -4.5 to -6.5 deg (C=1、DMF)