2-(4-Fluorophenyl)pyridine ≥98.0%

Leverantör: TCI

F0727-1G F0727-5G
TCIAF0727-1GEA 1350 SEK
TCIAF0727-1G TCIAF0727-5G
2-(4-Fluorophenyl)pyridine ≥98.0%
2-(4-Fluorophenyl)pyridine
Formel: C₁₁H₈FN
Kokpunkt: 119°C
MDL Number: MFCD06201382
CAS nummer: 58861-53-3
EINECS: 261-473-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=26406b801f496793fbe0529d02bc79a2d48745e1&vid=26406b801f496793fbe0529d02bc79a2d48745e1&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 38.0 to 42.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR