3-(4-Fluorophenyl)acrylaldehyde ≥95.0%
Leverantör: TCI EUROPE
Varning
Synonyms: 3-(4-Fluorophenyl)-2-propenal, 4-Fluorocinnamaldehyde, 3-(4-Fluorophenyl)acrylaldehyde
F0722-25G F0722-5G
TCIAF0722-25GEA 2025 SEK
TCIAF0722-25G TCIAF0722-5G
3-(4-Fluorophenyl)acrylaldehyde ≥95.0%
3-(4-Fluorophenyl)acrylaldehyde
Formel: C₉H₇FO
MW: 150,15 g/mol
Kokpunkt: 95°C
Storage Temperature: Kylskåp
MDL Number: MFCD03425895
CAS nummer: 24654-55-5
Order Now
Specifikation testresultat
Appearance White to Yellow to Green clear liquid
Purity (GC) min. 95.0 %