1-(4-Formylphenyl)pyrrolidine ≥98.0%
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
4-(1-Pyrrolidino)benzaldehyde, 4-(1-Pyrrolidinyl)benzaldehyde
F0679-1G
F0679-5G
TCIAF0679-1GEA
470
SEK
TCIAF0679-1G
TCIAF0679-5G
1-(4-Formylphenyl)pyrrolidine ≥98.0%
1-(4-Formylphenyl)pyrrolidine
Formel:
C₁₁H₁₃NO MW: 175,23 g/mol |
MDL Number:
MFCD00015469 CAS nummer: 51980-54-2 EINECS: 257-570-0 |
Specifikation testresultat
Appearance | Light yellow to Brown to Dark green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 82.0 to 86.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...