Flutamide ≥98.0%

Leverantör: TCI EUROPE
Fara

Synonyms: N-(4-Nitro-3-(trifluoromethyl)phenyl)isobutyramide, 4'-Nitro-3'-(trifluoromethyl)isobutyranilide, 2-Methyl-N-[4-nitro-3-(trifluoromethyl)phenyl]propionamide

F0663-25G F0663-5G
TCIAF0663-25GEA 1300 SEK
TCIAF0663-25G TCIAF0663-5G
Flutamide ≥98.0%
Flutamide
Formel: C₁₁H₁₁F₃N₂O₃
MW: 276,22 g/mol
MDL Number: MFCD00072009
CAS nummer: 13311-84-7
EINECS: 236-341-9
Merck Index: 14,04208

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 110.0 to 114.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR