(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-5-ureidopentanoic acid ≥98.0% (by HPLC, titration analysis)
Leverantör: TCI EUROPE
Synonyms:
N-Fmoc-L-citrulline, Nalpha-Fmoc-L-citrulline, Fmoc-Cit-OH
F0626-1G
F0626-5G
TCIAF0626-1GEA
245
SEK
TCIAF0626-1G
TCIAF0626-5G
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-5-ureidopentanoic acid ≥98.0% (by HPLC, titration analysis)
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-5-ureidopentanoic acid
Formel:
C₂₁H₂₃N₃O₅ MW: 397,4 g/mol |
MDL Number:
MFCD00151943 CAS nummer: 133174-15-9 |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Optical purity (LC) | min. 98.0 ee% |
Purity (Neutralization titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...