3-Fluorothiophene ≥98.0%
Leverantör: TCI EUROPE
Fara
F0512-200MG F0512-1G
TCIAF0512-200MGEA 2725 SEK
TCIAF0512-200MG TCIAF0512-1G
3-Fluorothiophene ≥98.0%
3-Fluorothiophene
Formel: C₄H₃FS
MW: 102,13 g/mol
Kokpunkt: 85°C
Storage Temperature: Frys
MDL Number: MFCD08275189
CAS nummer: 3093-88-7
UN: 1993
ADR: 3,II
Order Now
Specifikation testresultat
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %