4-Fluorocatechol ≥98.0%
Leverantör: TCI EUROPE
|
Fara
|
Synonyms:
4-Fluoro-1,2-dihydroxybenzene, 4-Fluoropyrocatechol
F0483-1G
F0483-5G
TCIAF0483-1GEA
1550
SEK
TCIAF0483-1G
TCIAF0483-5G
4-Fluorocatechol ≥98.0%
4-Fluorocatechol
Formel:
C₆H₅FO₂ MW: 128,10 g/mol |
MDL Number:
MFCD01317539 CAS nummer: 367-32-8 UN: 1759 ADR: 8,II |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 89.0 to 93.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...