N-Fmoc-L-threonine monohydrate ≥98.0% (by HPLC, titration analysis)

Leverantör: TCI EUROPE

Synonyms: N-(9-Fluorenylmethoxycarbonyl)-L-threonine monohydrate, Fmoc-Thr-OH monohydrate

F0455-1G F0455-25G F0455-5G
TCIAF0455-1GEA 215 SEK
TCIAF0455-1G TCIAF0455-25G TCIAF0455-5G
N-Fmoc-L-threonine monohydrate ≥98.0% (by HPLC, titration analysis)
N-Fmoc-L-threonine monohydrate
Formel: C₁₉H₁₉NO₅·H₂O
MW: 359,38 g/mol (341,36 anhy)
Storage Temperature: Frys
MDL Number: MFCD00792901
CAS nummer: 73731-37-0

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 % (calcd.on anh.substance)
Specific rotation [a]20/D -14.0 to -17.0 deg (C=1, DMF)
Solubility in Methanol very faint turbidity

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR