9,9-Bis[4-(2-hydroxyethoxy)phenyl]fluorene ≥98.0%

Leverantör: TCI
Varning

Synonyms: 4,4'-(9-Fluorenylidene)bis(2-phenoxyethanol), 2,2'-(((9H-Fluorene-9,9-diyl)bis(4,1-phenylene))bis(oxy))diethanol

F0447-25G F0447-5G
TCIAF0447-25GEA 910 SEK
TCIAF0447-25G TCIAF0447-5G
9,9-Bis[4-(2-hydroxyethoxy)phenyl]fluorene ≥98.0%
9,9-Bis[4-(2-hydroxyethoxy)phenyl]fluorene
Formel: C₂₉H₂₆O₄
Smältpunkt: 163°C
MDL Number: MFCD00216628
CAS nummer: 117344-32-8

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 160.0 to 166.0 deg-C
Solubility in hot Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR