5-Fluoro-2-methylbenzoxazole ≥97.0%
Leverantör: TCI EUROPE
Synonyms: 5-Fluoro-2-methylbenzo[d]oxazole
F0420-1G F0420-5G
TCIAF0420-1GEA 935 SEK
TCIAF0420-1G TCIAF0420-5G
5-Fluoro-2-methylbenzoxazole ≥97.0%
5-Fluoro-2-methylbenzoxazole
Formel: C₈H₆FNO
MW: 151,14 g/mol
Kokpunkt: 90°C
MDL Number: MFCD02093471
CAS nummer: 701-16-6
Order Now
Specifikation testresultat
Appearance White to Almost white clear liquid
Purity (GC) min. 97.0 %