4-Fluorophenyl chlorothionoformate ≥98.0%

Leverantör: TCI EUROPE
Fara

Synonyms: O-(4-Fluorophenyl) Chlorothioformate, Chlorothioformic Acid O-(4-Fluorophenyl) Ester

F0365-1G F0365-5G
TCIAF0365-1GEA 605 SEK
TCIAF0365-1G TCIAF0365-5G
4-Fluorophenyl chlorothionoformate ≥98.0%
4-Fluorophenyl chlorothionoformate
Formel: C₇H₄ClFOS
MW: 190,62 g/mol
Kokpunkt: 82°C
Flampunkt: 84°C
MDL Number: MFCD00134402
CAS nummer: 42908-73-6
UN: 1760
ADR: 8,III

Order Now

Specifikation testresultat

Appearance Light yellow to Yellow to Orange clear liquid
Purity (GC) min. 97.0 %
Purity (Argentmetric Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR