3-Furanmethanol ≥98.0%

Leverantör: TCI EUROPE
Varning

Synonyms: 3-Furyl Alcohol, 3-(Hydroxymethyl)furan

F0360-10G F0360-25G
TCIAF0360-10GEA 2525 SEK
TCIAF0360-10G TCIAF0360-25G
3-Furanmethanol ≥98.0%
3-Furanmethanol
Formel: C₅H₆O₂
MW: 98,10 g/mol
Kokpunkt: 100°C
Flampunkt: 38°C
MDL Number: MFCD00005352
CAS nummer: 4412-91-3
EINECS: 224-570-7
UN: 1987
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=74d57504d7759fb3a7f0e03223bd1eba3f9ac225&vid=74d57504d7759fb3a7f0e03223bd1eba3f9ac225&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR