Cyanomethyl formate ≥98.0% formylating reagent

Leverantör: TCI
Varning

Synonyms: Formic Acid Cyanomethyl Ester, Cyanomethylformate

F0356-25G F0356-5G
TCIAF0356-25GEA 4850 SEK
TCIAF0356-25G TCIAF0356-5G
Cyanomethyl formate ≥98.0% formylating reagent
Cyanomethyl formate
Formel: C₃H₃NO₂
Kokpunkt: 64°C
Flampunkt: 82°C
MDL Number: MFCD00661077
CAS nummer: 150760-95-5

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR