5-Formyl-2-furancarboxylic acid ≥98.0%
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
Furfural-5-carboxylic Acid, 5-Formylfuran-2-carboxylic acid, 5-Formyl-2-furoic Acid
F0338-1G
F0338-5G
TCIAF0338-1GEA
1350
SEK
TCIAF0338-1G
TCIAF0338-5G
5-Formyl-2-furancarboxylic acid ≥98.0%
5-Formyl-2-furancarboxylic acid
Formel:
C₆H₄O₄ MW: 140,09 g/mol |
MDL Number:
MFCD00020924 CAS nummer: 13529-17-4 |
Specifikation testresultat
Appearance | White to Yellow to Orange powder to crystal |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 206.0 to 212.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...