Ferroceneboronic acid ≥98.0% (contains varying amounts of Anhydride) cyclic boronating reagent for GC/MS

Leverantör: TCI EUROPE
Varning

F0280-1G F0280-100MG
TCIAF0280-1GEA 2625 SEK
TCIAF0280-1G TCIAF0280-100MG
Ferroceneboronic acid ≥98.0% (contains varying amounts of Anhydride) cyclic boronating reagent for GC/MS
Ferroceneboronic acid
Formel: C₁₀H₁₁BFeO₂
MW: 229,85 g/mol
MDL Number: MFCD00059074
CAS nummer: 12152-94-2
EINECS: 235-272-1

Order Now

Specifikation testresultat

Appearance White to Yellow to Orange powder to crystal
Purity (HPLC) min. 98.0 area%
Effect test of GC derivalization Effective as derivatization agent 3-Benzyloxy-1,2-propanediol

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR