Ethyl 2,4,5-trifluorobenzoylacetate ≥97.0%

Leverantör: TCI

Synonyms: (2,4,5-Trifluorobenzoyl)acetic acid ethyl ester, 3-Oxo-3-(2,4,5-trifluorophenyl)propanoic acid ethyl ester, Ethyl (2,4,5-Trifluorobenzoyl)acetate, Ethyl 2,4,5-trifluorobenzoylacetate, Ethyl 3-Oxo-3-(2,4,5-trifluorophenyl)propanoate

E1425-1G E1425-5G
TCIAE1425-1GEA 365 SEK
TCIAE1425-1G TCIAE1425-5G
Ethyl 2,4,5-trifluorobenzoylacetate ≥97.0%
Ethyl-2,4,5-trifluorobenzoylacetate
Formel: C₁₁H₉F₃O₃
MW: 246.19 g/mol
Kokpunkt: 92°C
MDL Number: MFCD00792431
CAS nummer: 98349-24-7

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 97.0 %
Melting point 65.0 to 69.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR