≥98.0% (contains 2-Ethyl-3,6-dimethylpyrazine)

Leverantör: TCI

E1424-100G E1424-25G
TCIAE1424-100GEA 2225 SEK
TCIAE1424-100G TCIAE1424-25G
≥98.0% (contains 2-Ethyl-3,6-dimethylpyrazine)
2-Ethyl-3,5-dimethylpyrazine
Formel: C₈H₁₂N₂
MW: 136.194 g/mol
Flampunkt: 69°C
MDL Number: MFCD00047392
CAS nummer: 27043-05-6
EINECS: 248-182-2

Order Now

Specifikation testresultat

Appearance White to Orange to Green clear liquid
Purity (GC) min. 98.0 % (total of isomers)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR