1-Ethyl-1H-pyrrole-2-carboxaldehyde ≥98.0%
Leverantör: TCI EUROPE
Synonyms: 1-Ethyl-1H-pyrrole-2-carbaldehyde, 2-Formyl-1-ethylpyrrole
E1416-1G E1416-5G
TCIAE1416-1GEA 725 SEK
TCIAE1416-1G TCIAE1416-5G
1-Ethyl-1H-pyrrole-2-carboxaldehyde ≥98.0%
1-Ethyl-1H-pyrrole-2-carboxaldehyde
Formel: C₇H₉NO
MW: 123,16 g/mol
Kokpunkt: 75°C
MDL Number: MFCD11909543
CAS nummer: 2167-14-8
Order Now
Specifikation testresultat
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %