2-Ethoxy-4-formylphenyl acetate ≥98.0%
Leverantör: TCI
Varning
Synonyms: Ethylvanillin acetate
E1415-25G E1415-5G
TCIAE1415-25GEA 910 SEK
TCIAE1415-25G TCIAE1415-5G
2-Ethoxy-4-formylphenyl acetate ≥98.0%
2-Ethoxy-4-formylphenyl acetate
Formel: C₁₁H₁₂O₄
Storage Temperature: Kylskåp
MDL Number: MFCD01666413
CAS nummer: 72207-94-4
Order Now
Specifikation testresultat
Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 43.0 to 47.0 deg-C