3-(Ethoxydimethylsilyl)propyl methacrylate ≥95.0% stabilised

Leverantör: TCI EUROPE
Varning

Synonyms: Methacrylic acid 3-(Ethoxydimethylsilyl)propyl ester, (3-Methacryloyloxy)propyldimethylethoxysilane

Stabilization: Stabilised med BHT/Ionol

E1400-1G E1400-5G
TCIAE1400-1GEA 495 SEK
TCIAE1400-1G TCIAE1400-5G
3-(Ethoxydimethylsilyl)propyl methacrylate ≥95.0% stabilised
3-(Ethoxydimethylsilyl)propyl methacrylate
Formel: C₁₁H₂₂O₃Si
MW: 230,38 g/mol
Storage Temperature: Kylskåp
MDL Number: MFCD00053877
CAS nummer: 13731-98-1

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR