4-Ethoxyphenyl trans-4-butylcyclohexanecarboxylate ≥98.0%

Leverantör: TCI

Synonyms: Trans-4-ethoxyphenyl 4-butylcyclohexanecarboxylate, trans-4-Butylcyclohexanecarboxylic Acid 4-Ethoxyphenyl Ester

E1396-25G E1396-100G
TCIAE1396-25GEA 1550 SEK
TCIAE1396-25G TCIAE1396-100G
4-Ethoxyphenyl trans-4-butylcyclohexanecarboxylate ≥98.0%
4-Ethoxyphenyl trans-4-butylcyclohexanecarboxylate
Formel: C₁₉H₂₈O₃
Storage Temperature: Frys
MDL Number: MFCD01941073
CAS nummer: 67589-47-3
EINECS: 266-745-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3dd82990b60ddc860fa54a00c73b8279c7f073f5&vid=ad6926987df817fa689329d1131179120c6d7a4f&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Mesomorphic range 47.0 to 82.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR