(3-Ethyloxetan-3-yl)methyl methacrylate ≥98.0% stabilised
Leverantör: TCI
Synonyms:
Methacrylic acid (3-Ethyloxetan-3-yl)methyl ester
Stabilization:
Stabilised with 4-methoxyphenol (MEHQ)
E1379-1G
E1379-5G
TCIAE1379-1GEA
1100
SEK
TCIAE1379-1G
TCIAE1379-5G
(3-Ethyloxetan-3-yl)methyl methacrylate ≥98.0% stabilised
(3-Ethyloxetan-3-yl)methyl methacrylate
Formel:
C₁₀H₁₆O₃ MW: 184.24 g/mol |
MDL Number:
MFCD16039335 CAS nummer: 37674-57-0 |
Specifikation testresultat
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...