Epothilone B ≥95.0%
Leverantör: TCI
Synonyms:
(1S,3S,7S,10R,11S,12S,16R)-7,11-Dihydroxy-8,8,10,12,16-pentamethyl-3-((E)-1-(2-methylthiazol-4-yl)prop-1-en-2-yl)-4,17-dioxabicyclo[14.1.0]heptadecane-5,9-dione
E1378-25MG
E1378-5MG
TCIAE1378-25MGEA
5150
SEK
TCIAE1378-25MG
TCIAE1378-5MG
Epothilone B ≥95.0%
Epothilone B
Formel:
C₂₇H₄₁NO₆S Smältpunkt: 94°C Storage Temperature: Frys |
MDL Number:
MFCD02101921 CAS nummer: 152044-54-7 Merck Index: 14,03628 |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 95.0 area% |
Specific rotation [a]20/D | -40.0 to -44.0 deg (C=0.2, methanol) |
Elemental analysis (Carbon) | 62.50 to 65.00 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...