4-Ethynylbenzyl alcohol ≥98.0%

Leverantör: TCI

E1377-1G E1377-5G
TCIAE1377-1GEA 935 SEK
TCIAE1377-1G TCIAE1377-5G
4-Ethynylbenzyl alcohol ≥98.0%
4-Ethynylbenzyl alcohol
Formel: C₉H₈O
Kokpunkt: 66°C
Storage Temperature: Kylskåp
MDL Number: MFCD05664205
CAS nummer: 10602-04-7

Order Now

Specifikation testresultat

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 38.0 to 42.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR