Elacridar ≥98.0%
Leverantör: TCI EUROPE
Synonyms:
N-[4-[2-(3,4-Dihydro-6,7-dimethoxy-2(1H)-isoquinolinyl)ethyl]phenyl]-9,10-dihydro-5-methoxy-9-oxo-4-acridinecarboxamide, N-(4-(2-(6,7-Dimethoxy-3,4-dihydroisoquinolin-2(1H)-yl)ethyl)phenyl)-5-methoxy-9-oxo-9,10-dihydroacridine-4-carboxamide
E1371-100MG
E1371-25MG
TCIAE1371-100MGEA
4850
SEK
TCIAE1371-100MG
TCIAE1371-25MG
Elacridar ≥98.0%
Elacridar
Formel:
C₃₄H₃₃N₃O₅ MW: 563,654 g/mol Storage Temperature: Frys |
MDL Number:
MFCD00912604 CAS nummer: 143664-11-3 |
Specifikation testresultat
Appearance | White to Yellow to Green powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Elemental analysis (Nitrogen) | 7.00 to 8.00 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...