trans-1-Ethoxy-4-(4-n-propycyclohexyl)benzene ≥98.0%
Leverantör: TCI EUROPE
Synonyms:
1-Ethoxy-4-(trans-4-propylcyclohexyl)benzene
E1369-25G
E1369-5G
TCIAE1369-25GEA
1725
SEK
TCIAE1369-25G
TCIAE1369-5G
trans-1-Ethoxy-4-(4-n-propycyclohexyl)benzene ≥98.0%
trans-1-Ethoxy-4-(4-n-propycyclohexyl)benzene
Formel:
C₁₇H₂₆O MW: 246,394 g/mol |
MDL Number:
MFCD06658195 CAS nummer: 80944-44-1 EINECS: 279-633-1 |
Specifikation testresultat
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 40.0 to 44.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...