(4-Ethynylphenyl)trimethylsilane ≥98.0%
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
1-Ethynyl-4-(trimethylsilyl)benzene, 4-(Trimethylsilyl)phenylacetylene
E1362-1G
E1362-5G
TCIAE1362-1GEA
1350
SEK
TCIAE1362-1G
TCIAE1362-5G
(4-Ethynylphenyl)trimethylsilane ≥98.0%
(4-Ethynylphenyl)trimethylsilane
Formel:
C₁₁H₁₄Si MW: 174,32 g/mol Kokpunkt: 75°C Storage Temperature: Kylskåp |
MDL Number:
MFCD26394823 CAS nummer: 16116-92-0 |
Specifikation testresultat
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...