4-(2-Ethylhexyl)-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)thiophene ≥97.0%

Leverantör: TCI EUROPE

Synonyms: 4-(2-Ethylhexyl)-2-thiopheneboronic acid Pinacol ester, 2-[4-(2-Ethylhexyl)-2-thienyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

E1355-1G E1355-5G
TCIAE1355-1GEA 1750 SEK
TCIAE1355-1G TCIAE1355-5G
4-(2-Ethylhexyl)-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)thiophene ≥97.0%
4-(2-Ethylhexyl)-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)thiophene
Formel: C₁₈H₃₁BO₂S
MW: 322,31 g/mol
Storage Temperature: Kylskåp
CAS nummer: 1224430-50-5

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR