1-Ethynyl-3,5-difluorobenzene ≥98.0%
Leverantör: TCI EUROPE
Varning
Synonyms: 3,5-Difluorophenylacetylene
E1352-1G E1352-5G
TCIAE1352-1GEA 990 SEK
TCIAE1352-1G TCIAE1352-5G
1-Ethynyl-3,5-difluorobenzene ≥98.0%
1-Ethynyl-3,5-difluorobenzene
Formel: C₈H₄F₂
MW: 138,12 g/mol
Flampunkt: 33°C
Storage Temperature: Kylskåp
MDL Number: MFCD04039982
CAS nummer: 151361-87-4
UN: 1993
ADR: 3,III
Order Now
Specifikation testresultat
Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 98.0 %