2,2-Difluoropropionic acid ethyl ester ≥95.0%
Leverantör: TCI
|
Fara
|
Synonyms:
Ethyl 2,2-Difluoropropionate, 2,2-Difluoropropionic acid ethyl ester
E1350-1G
E1350-5G
TCIAE1350-1GEA
365
SEK
TCIAE1350-1G
TCIAE1350-5G
2,2-Difluoropropionic acid ethyl ester ≥95.0%
2,2-Difluoropropionic acid ethyl ester
Formel:
C₅H₈F₂O₂ Kokpunkt: 115°C |
MDL Number:
MFCD06657970 CAS nummer: 28781-85-3 UN: 3272 ADR: 3,II |
Specifikation testresultat
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...