1-Ethoxy-4-[2-(4-methylphenyl)ethynyl]benzene ≥98.0%

Leverantör: TCI EUROPE

Synonyms: 1-Ethoxy-4-[2-(4-methylphenyl)ethynyl]benzene, 1-Ethoxy-4-(p-tolylethynyl)benzene

E1342-1G E1342-5G
TCIAE1342-1GEA 405 SEK
TCIAE1342-1G TCIAE1342-5G
1-Ethoxy-4-[2-(4-methylphenyl)ethynyl]benzene ≥98.0%
1-Ethoxy-4-[2-(4-methylphenyl)ethynyl]benzene
Formel: C₁₇H₁₆O
MW: 236,314 g/mol
MDL Number: MFCD01817758
CAS nummer: 116903-46-9

Order Now

Specifikation testresultat

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 106.0 to 110.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR