2-(Ethoxycarbonyl)phenylboronic acid pinacol ester ≥98.0%
Leverantör: TCI EUROPE
Synonyms:
2-Ethoxycarbonylphenylboronic acid pinacol ester, 2-(Ethoxycarbonyl)benzeneboronic acid pinacol ester, Ethyl-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate, Ethyl 2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate
E1311-1G
E1311-5G
TCIAE1311-1GEA
1025
SEK
TCIAE1311-1G
TCIAE1311-5G
2-(Ethoxycarbonyl)phenylboronic acid pinacol ester ≥98.0%
2-(Ethoxycarbonyl)phenylboronic acid pinacol ester
Formel:
C₁₅H₂₁BO₄ MW: 276,136 g/mol |
MDL Number:
MFCD03453660 CAS nummer: 269409-99-6 |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 97.0 % |
Melting point | 63.0 to 68.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...