Benzarone 98,0 GC_ANALYSMETOD
Leverantör: TCI
|
Varning
|
Synonyms:
(2-Ethylbenzofuran-3-yl)(4-hydroxyphenyl)methanone, 2-Ethyl-3-(4-hydroxybenzoyl)benzofuran
E1289-1G
E1289-5G
TCIAE1289-1GEA
1450
SEK
TCIAE1289-1G
TCIAE1289-5G
Benzarone 98,0 GC_ANALYSMETOD
Benzarone
Formel:
C₁₇H₁₄O₃ MW: 266.30 g/mol Storage Temperature: Kylskåp |
MDL Number:
MFCD00867417 CAS nummer: 1477-19-6 EINECS: 216-026-2 |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 124.0 to 128.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...