Ethyl-2-chloro-6-methylisonicotinate ≥98.0%
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
Ethyl 2-chloro-6-methylpyridine-4-carboxylate, Ethyl 2-chloro-6-methylisonicotinate
E1262-1G
E1262-5G
TCIAE1262-1GEA
645
SEK
TCIAE1262-1G
TCIAE1262-5G
Ethyl-2-chloro-6-methylisonicotinate ≥98.0%
Ethyl-2-chloro-6-methylisonicotinate
Formel:
C₉H₁₀ClNO₂ MW: 199,64 g/mol Kokpunkt: 107°C |
MDL Number:
MFCD09842151 CAS nummer: 3998-88-7 |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 65.0 to 69.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...