trans-4-Ethyl-4'-(4-n-pentylcyclohexyl)biphenyl ≥98.0%

Leverantör: TCI EUROPE

Synonyms: 4-Ethyl-4'-(trans-4-pentylcyclohexyl)biphenyl, 4-Ethyl-4'-(trans-4-pentylcyclohexyl)-1,1'-biphenyl

E1254-1G E1254-5G
TCIAE1254-1GEA 565 SEK
TCIAE1254-1G TCIAE1254-5G
trans-4-Ethyl-4'-(4-n-pentylcyclohexyl)biphenyl ≥98.0%
trans-4-Ethyl-4'-(4-n-pentylcyclohexyl)biphenyl
Formel: C₂₅H₃₄
MW: 334,547 g/mol
MDL Number: MFCD01941072
CAS nummer: 79709-85-6
EINECS: 279-240-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0bb749931fb2a64b7fc082351592a91e6c93b42c&vid=0bb749931fb2a64b7fc082351592a91e6c93b42c&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Mesomorphic range 144.0 to 167.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR