Ethyl-3-hydroxy-2-naphthoate ≥98.0%

Leverantör: TCI EUROPE

Synonyms: 3-Hydroxy-2-naphthoic acid ethyl ester, Ethyl 3-hydroxy-2-naphthoate

E1249-1G E1249-5G
TCIAE1249-1GEA 1025 SEK
TCIAE1249-1G TCIAE1249-5G
Ethyl-3-hydroxy-2-naphthoate ≥98.0%
Ethyl-3-hydroxy-2-naphthoate
Formel: C₁₃H₁₂O₃
MW: 216,2326 g/mol
Kokpunkt: 291°C
MDL Number: MFCD00220608
CAS nummer: 7163-25-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ffb3323bdc8471952c11768d9be6540b78631299&vid=ffb3323bdc8471952c11768d9be6540b78631299&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 79.0 to 83.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR