Ethyl ethoxyacetate ≥95.0%
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
Ethyl 2-ethoxyacetate, Ethoxyacetic acid ethyl ester
E1237-5G
E1237-25G
TCIAE1237-5GEA
485
SEK
TCIAE1237-5G
TCIAE1237-25G
Ethyl ethoxyacetate ≥95.0%
Ethyl ethoxyacetate
Formel:
C₆H₁₂O₃ MW: 132,16 g/mol Kokpunkt: 158°C Flampunkt: 50°C |
MDL Number:
MFCD00040492 CAS nummer: 817-95-8 EINECS: 212-447-0 UN: 3272 ADR: 3,III |
Specifikation testresultat
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 95.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...