4-Ethoxy-3-(trifluoromethyl)phenylboronic acid (contains varying amounts of Anhydride)

Leverantör: TCI EUROPE
Varning

Synonyms: 4-Ethoxy-3-(trifluoromethyl)benzeneboronic acid, 4-Ethoxy-3-trifluoromethylphenylboronic acid

E1236-1G E1236-200MG
TCIAE1236-1GEA 2725 SEK
TCIAE1236-1G TCIAE1236-200MG
4-Ethoxy-3-(trifluoromethyl)phenylboronic acid (contains varying amounts of Anhydride)
4-Ethoxy-3-(trifluoromethyl)phenylboronic acid
Formel: C₉H₁₀BF₃O₃
MW: 233,98 g/mol
MDL Number: MFCD07363789
CAS nummer: 871329-83-8

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) 97.0 to 108.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR