Ethyl (Benzyloxy)acetate ≥98.0%
Leverantör: TCI EUROPE
Synonyms: Ethyl 2-(benzyloxy)acetate, (Benzyloxy)acetic acid Ethyl ester
E1225-1G E1225-5G
TCIAE1225-1GEA 510 SEK
TCIAE1225-1G TCIAE1225-5G
Ethyl (Benzyloxy)acetate ≥98.0%
Ethyl (Benzyloxy)acetate
Formel: C₁₁H₁₄O₃
MW: 194,23 g/mol
Kokpunkt: 287°C
Storage Temperature: Kylskåp
MDL Number: MFCD06204614
CAS nummer: 32122-09-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a5ed23c97a6105e75743ba90feaff6b3aa2344c0&vid=a5ed23c97a6105e75743ba90feaff6b3aa2344c0&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %
Acid Value max. 2.0