1-Ethynyl-4-nitrobenzene ≥98.0%

Leverantör: TCI
Varning

Synonyms: 4-Nitrophenylacetylene

E1223-1G E1223-5G
TCIAE1223-1GEA 870 SEK
TCIAE1223-1G TCIAE1223-5G
1-Ethynyl-4-nitrobenzene ≥98.0%
1-Ethynyl-4-nitrobenzene
Formel: C₈H₅NO₂
MW: 147.13 g/mol
MDL Number: MFCD00024794
CAS nummer: 937-31-5

Order Now

Specifikation testresultat

Purity (GC) min. 98.0 %
Melting point 149.0 to 153.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR