5-(Ethoxycarbonyl)-2-fluorophenylboronic acid (contains varying amounts of Anhydride)

Leverantör: TCI EUROPE
Varning

Synonyms: (5-(Ethoxycarbonyl)-2-fluorophenyl)boronic acid, 5-Ethoxycarbonyl-2-fluorobenzeneboronic acid

E1209-1G E1209-200MG
TCIAE1209-1GEA 1425 SEK
TCIAE1209-1G TCIAE1209-200MG
5-(Ethoxycarbonyl)-2-fluorophenylboronic acid (contains varying amounts of Anhydride)
5-(Ethoxycarbonyl)-2-fluorophenylboronic acid
Formel: C₉H₁₀BFO₄
MW: 211,98 g/mol
MDL Number: MFCD08235094
CAS nummer: 874219-60-0

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) 98.0 to 109.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR