Ethyl 2-Iodo-2-methylpropionate ≥94.0%

Leverantör: TCI EUROPE
Varning

Synonyms: 2-Iodo-2-methylpropionic acid Ethyl ester

E1199-5G E1199-25G
TCIAE1199-5GEA 630 SEK
TCIAE1199-5G TCIAE1199-25G
Ethyl 2-Iodo-2-methylpropionate ≥94.0%
Ethyl 2-Iodo-2-methylpropionate
Formel: C₆H₁₁IO₂
MW: 242,06 g/mol
Storage Temperature: Frys
MDL Number: MFCD29089363
CAS nummer: 7425-55-0

Order Now

Specifikation testresultat

Appearance Light yellow to Brown clear liquid
Purity (GC) min. 94.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR