2,3-Difluoro-4-ethoxyphenylboronic acid (contains varying amounts of Anhydride)

Leverantör: TCI EUROPE
Varning

Synonyms: 2,3-Difluoro-4-ethoxybenzeneboronic acid, 4-Ethoxy-2,3-difluorobenzeneboronic acid, 4-Ethoxy-2,3-difluorophenylboronic acid, (4-Ethoxy-2,3-difluorophenyl)boronic acid

E1192-1G E1192-5G
TCIAE1192-1GEA 310 SEK
TCIAE1192-1G TCIAE1192-5G
2,3-Difluoro-4-ethoxyphenylboronic acid (contains varying amounts of Anhydride)
2,3-Difluoro-4-ethoxyphenylboronic acid
Formel: C₈H₉BF₂O₃
MW: 201,96 g/mol
MDL Number: MFCD09258743
CAS nummer: 212386-71-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6bf7a10621c6a5b4ab5af5bb389a8adef24b8ed6&vid=6bf7a10621c6a5b4ab5af5bb389a8adef24b8ed6&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) 97.0 to 110.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR