3-(Ethoxycarbonyl)-4-fluorophenylboronic acid (contains varying amounts of Anhydride)

Leverantör: TCI
Varning

Synonyms: 3-Ethoxycarbonyl-4-fluorophenylboronic acid, (3-Ethoxycarbonyl-4-fluorophenyl)boronic acid

E1187-1G E1187-5G
TCIAE1187-1GEA 855 SEK
TCIAE1187-1G TCIAE1187-5G
3-(Ethoxycarbonyl)-4-fluorophenylboronic acid (contains varying amounts of Anhydride)
3-(Ethoxycarbonyl)-4-fluorophenylboronic acid
Formel: C₉H₁₀BFO₄
MW: 211.9827 g/mol
MDL Number: MFCD08061836
CAS nummer: 874219-36-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b2e58050615dc81bfe9130374cc4fb6f9c8fdfdc&vid=b2e58050615dc81bfe9130374cc4fb6f9c8fdfdc&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) 98.0 to 109.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR