Ethyl 3,4-Bis(2-methoxyethoxy)benzoate ≥98.0%

Leverantör: TCI

E1186-1G E1186-5G
TCIAE1186-1GEA 365 SEK
TCIAE1186-1G TCIAE1186-5G
Ethyl 3,4-Bis(2-methoxyethoxy)benzoate ≥98.0%
Ethyl 3,4-Bis(2-methoxyethoxy)benzoate
Formel: C₁₅H₂₂O₆
MDL Number: MFCD01935284
CAS nummer: 183322-16-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1954ff4995541de5917269d0979d87dafa31f58d&vid=1954ff4995541de5917269d0979d87dafa31f58d&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 54.0 to 58.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR