2-Ethyl-6-methylphenol ≥98.0%

Leverantör: TCI
Varning

E1185-1G E1185-5G
TCIAE1185-1GEA 405 SEK
TCIAE1185-1G TCIAE1185-5G
2-Ethyl-6-methylphenol ≥98.0%
2-Ethyl-6-methylphenol
Formel: C₉H₁₂O
Kokpunkt: 214°C
MDL Number: MFCD00962292
CAS nummer: 1687-64-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4c682fa516a5d03e7bb571a6bcaad13ad2b32f41&vid=4c682fa516a5d03e7bb571a6bcaad13ad2b32f41&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Light yellow to Brown clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR