Ethyl-2-((dimethylamino)methylene)-3-oxobutanoate ≥95.0% (by GC, titration analysis)
Leverantör: TCI EUROPE
Synonyms:
Ethyl 2-((dimethylamino)methylene)-3-oxobutanoate, Ethyl 2-acetyl-3-(dimethylamino)acrylate
E1179-5G
E1179-25G
TCIAE1179-5GEA
535
SEK
TCIAE1179-5G
TCIAE1179-25G
Ethyl-2-((dimethylamino)methylene)-3-oxobutanoate ≥95.0% (by GC, titration analysis)
Ethyl-2-((dimethylamino)methylene)-3-oxobutanoate
Formel:
C₉H₁₅NO₃ MW: 185,22 g/mol Kokpunkt: 150°C |
MDL Number:
MFCD00087563 CAS nummer: 51145-57-4 |
Specifikation testresultat
Appearance | Light yellow to Brown clear liquid |
Purity (GC) | min. 95.0 % |
Purity (Nonaqueous Titration) | min. 95.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...